Assay Details
| Gene Symbol: Tspan12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173007.3 | NP_766595.1 | 7 - 8 | 808 | 77 | |
| GenBank mRNA | AK033554.1 | - | 6 - 7 | 643 | 77 | |
| AK050737.1 | - | 7 - 8 | 809 | 77 | ||
| AK199414.1 | - | 1 - 2 | 54 | 77 | ||
| BC068240.1 | - | 6 - 7 | 670 | 3674585 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tspan12
Genomic Map