Assay Details
Gene Symbol: Ssc5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173008.2 | NP_766596.1 | 1 - 2 | 137 | 75 | |
GenBank mRNA | AK079906.1 | - | 1 - 2 | 138 | 75 | |
AK165666.1 | - | 1 - 2 | 199 | 75 | ||
BC141044.1 | - | 1 - 2 | 135 | 9055656 | 75 | |
EU850434.1 | - | 1 - 2 | 138 | 75 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK079906.1,AK165666.1,BC141044.1,EU850434.1,NM_173008.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
