Assay Details
| Gene Symbol: Lpcat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173014.1 | NP_766602.1 | 10 - 11 | 1164 | 71 | |
| GenBank mRNA | AB244716.1 | - | 10 - 11 | 1059 | 71 | |
| AK039431.1 | - | 10 - 11 | 1164 | 71 | ||
| BC144805.1 | - | 9 - 10 | 1058 | 71 | ||
| BC144806.1 | - | 8 - 9 | 999 | 9053354 | 71 | |
| BC150666.1 | - | 8 - 9 | 999 | 9087577 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lpcat2
Genomic Map