Assay Details
| Gene Symbol: Lpcat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173014.1 | NP_766602.1 | 11 - 12 | 1320 | 86 | |
| GenBank mRNA | AB244716.1 | - | 11 - 12 | 1215 | 86 | |
| AK039431.1 | - | 11 - 12 | 1320 | 86 | ||
| BC144805.1 | - | 10 - 11 | 1214 | 86 | ||
| BC144806.1 | - | 9 - 10 | 1155 | 9053354 | 86 | |
| BC150666.1 | - | 9 - 10 | 1155 | 9087577 | 86 | |
More Information
Set Membership: |
|

Lpcat2
Genomic Map