Assay Details
| Gene Symbol: Mon1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048143.2 | NP_001041608.1 | 2 - 3 | 286 | 74 | |
| NM_173015.3 | NP_766603.1 | 1 - 2 | 476 | 74 | ||
| GenBank mRNA | AK030288.1 | - | 1 - 2 | 442 | 74 | |
| AK157390.1 | - | 2 - 3 | 286 | 74 | ||
| AK173047.1 | - | 1 - 2 | 148 | 74 | ||
| BC062639.1 | - | 1 - 2 | 152 | 74 | ||
| CA320533.1 | - | 1 - 2 | 66 | 74 | ||
| CJ089261.1 | - | 2 - 3 | 287 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26179657 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26179658 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26179659 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26179660 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26179661 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2076471 | AK173047.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2076471 | BC062639.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2076471 | NM_001048143.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2076471 | NM_173015.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2076471 | AK017167.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 32300 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 634339 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649097 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 657280 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 645635 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 76487 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2076471 | AK173047.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2076471 | BC062639.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2076471 | NM_001048143.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2076471 | NM_173015.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2076471 | AK017167.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179648 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179657 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179682 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179644 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179671 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179636 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179638 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179632 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26179631 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01832134 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06647277 | - | - |
| Whole Mouse Genome G4122A | A_52_P56606 | AK173047.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56606 | BC062639.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56606 | NM_001048143.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56606 | NM_173015.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56606 | AK017167.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P143757 | AK157390.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P143757 | AK173047.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P143757 | BC062639.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P143757 | BQ831297.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P143757 | NM_001048143.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102074 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06647277 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07045111 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07045116 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07045112 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07045123 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26179657 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26179659 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26179658 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26179661 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26179660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26179658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07208506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26179627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26179656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07031251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26179624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26179648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26179647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26179618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26179662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26179626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07125540 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26179649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26179639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26179645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07045123 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26179652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26179621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07287873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26179636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26179620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06951523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26179613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26179660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26179615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07125536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26179640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26179634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26179623 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10575673 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575665 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575671 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575669 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575668 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575664 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575663 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575670 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575666 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575667 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432685_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428711_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428712_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10575662 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10575672 | - | - |
| Mouse Genome 430 2.0 Array | 1428711_at | - | - |
| Mouse Genome 430 2.0 Array | 1428712_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1432685_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428712_at | - | - |
| Mouse Genome 430 2.0 Array | 1432685_at | - | - |
| Mouse Genome 430 2.0 Array | 1428711_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 270096_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 270096_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800011130.mm.1 | NM_173015.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011130.mm.1 | TC0800001357.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800011133.mm.1 | NM_001048143.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011133.mm.1 | NM_173015.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011133.mm.1 | TC0800001357.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800011129.mm.1 | NM_001048143.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011129.mm.1 | TC0800001357.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800011132.mm.1 | NM_001048143.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011132.mm.1 | NM_173015.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011132.mm.1 | TC0800001357.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428711_PM_at | - | - |
| Mouse Expression Set 430 | 1428711_at | - | - |
| Mouse Expression Set 430 | 1428712_at | - | - |
| Mouse Expression Set 430 | 1428712_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428712_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432685_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432685_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428712_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428711_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428711_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 775965 | - | - |
| ABI Mouse Genome Survey Microarray | 829355 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5890037 | - | - |
| MouseWG-6 v2 Expression BeadChip | 102480270 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5890037 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3520373 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102480270 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003520373 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005890037 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1219118 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2594558 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1219118 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2594558 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1219281 | - | - |
| mouse-6 v1.1 (Illumina) | scl00270096.1_196-S | - | - |
| mouse-6 v1.1 (Illumina) | scl33304.1.1_129-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0270096.5_118-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102480270 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5890037 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3520373 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0270096.5_118-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00270096.1_196-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3520373 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5890037 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102480270 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3053203 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2594558 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1219118 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1219118 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2594558 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3053203 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00270096.1_196-S | - | - |

Mon1b
Genomic Map