Assay Details
| Gene Symbol: Galnt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173030.2 | NP_766618.2 | 5 - 6 | 989 | 105 | |
| GenBank mRNA | AB082928.1 | - | 3 - 4 | 477 | 105 | |
| AK038387.1 | - | 5 - 6 | 990 | 105 | ||
| AK045417.1 | - | 5 - 6 | 990 | 105 | ||
| BC131651.1 | - | 5 - 6 | 668 | 40110517 | 105 | |
| BC131652.1 | - | 4 - 5 | 699 | 40110518 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galnt13
Genomic Map