Assay Details
| Gene Symbol: Rbbp8nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173031.3 | NP_766619.2 | 5 - 6 | 486 | 83 | |
| GenBank mRNA | AK028610.1 | - | 5 - 6 | 488 | 83 | |
| BC066135.1 | - | 5 - 6 | 442 | 83 | ||
| BC150940.1 | - | 5 - 6 | 441 | 83 | ||
| BC172093.1 | - | 5 - 6 | 441 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbbp8nl
Genomic Map