Assay Details
| Gene Symbol: Tbck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK034533.1 | - | 9 - 10 | 1022 | 61 | |
| AK052841.1 | - | 9 - 10 | 1034 | 61 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Tbck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK034533.1 | - | 9 - 10 | 1022 | 61 | |
| AK052841.1 | - | 9 - 10 | 1034 | 61 | ||
Set Membership: |
|