Assay Details
| Gene Symbol: Tbcel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173038.3 | NP_766626.1 | 3 - 4 | 457 | 68 | |
| GenBank mRNA | AK052261.1 | - | 3 - 4 | 426 | 68 | |
| AK077017.1 | - | 3 - 4 | 459 | 68 | ||
| AK084854.1 | - | 3 - 4 | 470 | 68 | ||
| BC047462.1 | - | 3 - 4 | 324 | 68 | ||
| BC139386.1 | - | 3 - 4 | 455 | 8862408 | 68 | |
| BC139387.1 | - | 3 - 4 | 455 | 8862409 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbcel
Genomic Map