Assay Details
| Gene Symbol: Tbcel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173038.3 | NP_766626.1 | 4 - 5 | 633 | 98 | |
| GenBank mRNA | AK052261.1 | - | 4 - 5 | 602 | 98 | |
| AK077017.1 | - | 4 - 5 | 635 | 98 | ||
| AK212783.1 | - | 1 - 2 | 131 | 98 | ||
| BC047462.1 | - | 4 - 5 | 500 | 98 | ||
| BC139386.1 | - | 4 - 5 | 631 | 8862408 | 98 | |
| BC139387.1 | - | 4 - 5 | 631 | 8862409 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbcel
Genomic Map