Assay Details
| Gene Symbol: Zc2hc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173181.3 | NP_775273.1 | 2 - 3 | 173 | 121 | |
| GenBank mRNA | AK014202.1 | - | 2 - 3 | 109 | 121 | |
| AK083941.1 | - | 2 - 3 | 117 | 121 | ||
| AK085187.1 | - | 2 - 3 | 110 | 121 | ||
| AK170609.1 | - | 2 - 3 | 114 | 121 | ||
| BC044809.1 | - | 2 - 3 | 95 | 121 | ||
| BC087731.1 | - | 137 | 6850296 | 121 | ||
| BY131077.1 | - | 2 - 3 | 174 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zc2hc1a
Genomic Map