Assay Details
| Gene Symbol: H6pd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291004.1 | NP_001277933.1 | 1 - 2 | 202 | 68 | |
| GenBank mRNA | AK159373.1 | - | 1 - 2 | 144 | 68 | |
| AK172249.1 | - | 1 - 2 | 69 | |||
| BC042677.1 | - | 1 - 2 | 96 | 4195793 | 68 | |
| BY149778.1 | - | 1 - 2 | 143 | 68 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK159373.1,BC042677.1,NM_001291004.1 transcripts.
Set Membership: |
|

H6pd
Genomic Map