Assay Details
| Gene Symbol: Trp53bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173378.2 | NP_775554.2 | 4 - 5 | 630 | 54 | |
| GenBank mRNA | BC042874.1 | - | 3 - 4 | 232 | 54 | |
| BQ180182.1 | - | 4 - 5 | 615 | 54 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trp53bp2
Genomic Map