Assay Details
| Gene Symbol: Trp53bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173378.2 | NP_775554.2 | 14 - 15 | 3112 | 86 | |
| GenBank mRNA | AK134556.1 | - | 7 - 8 | 2082 | 86 | |
| BC030894.1 | - | 6 - 7 | 1741 | 86 | ||
| BC042874.1 | - | 13 - 14 | 2714 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trp53bp2
Genomic Map