Assay Details
| Gene Symbol: Rab8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173413.3 | NP_775589.1 | 3 - 4 | 314 | 122 | |
| GenBank mRNA | AB232601.1 | - | 3 - 4 | 242 | 122 | |
| AK051218.1 | - | 3 - 4 | 264 | 122 | ||
| AK084650.1 | - | 3 - 4 | 264 | 122 | ||
| AK155621.1 | - | 3 - 4 | 264 | 122 | ||
| BC059208.1 | - | 3 - 4 | 296 | 6405462 | 122 | |
| BC157974.1 | - | 3 - 4 | 262 | 9007296 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab8b
Genomic Map