Assay Details
| Gene Symbol: Nbeal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173444.2 | NP_775620.2 | 51 - 52 | 7847 | 92 | |
| GenBank mRNA | AK041108.1 | - | 11 - 12 | 1185 | 92 | |
| BC055813.1 | - | 14 - 15 | 1576 | 92 | ||
| BC060141.1 | - | 27 - 28 | 3978 | 92 | ||
More Information
Set Membership: |
|

Nbeal1
Genomic Map