Assay Details
| Gene Symbol: Galnt18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173739.3 | NP_776100.2 | 1 | 46 | 54 | |
| GenBank mRNA | AK084671.1 | - | 1 | 51 | 54 | |
| BC024988.1 | - | 1 | 46 | 5044626 | 54 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC024988.1,NM_173739.3 transcripts.
Set Membership: |
|

Galnt18
Genomic Map