Assay Details
| Gene Symbol: Gpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173866.3 | NP_776291.1 | 3 - 4 | 433 | 66 | |
| GenBank mRNA | AK031958.1 | - | 3 - 4 | 445 | 66 | |
| AK033424.1 | - | 3 - 4 | 434 | 66 | ||
| AK075894.1 | - | 3 - 4 | 452 | 66 | ||
| AK076250.1 | - | 3 - 4 | 451 | 66 | ||
| AK082030.1 | - | 3 - 4 | 476 | 66 | ||
| BC034219.1 | - | 3 - 4 | 465 | 5065322 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpt2
Genomic Map