Assay Details
| Gene Symbol: Disc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174854.2 | NP_777279.2 | 8 - 9 | 1817 | 69 | |
| GenBank mRNA | AF513723.1 | - | 8 - 9 | 1814 | 69 | |
| AY177673.1 | - | 8 - 9 | 1782 | 69 | ||
| AY320287.1 | - | 9 - 10 | 1899 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Disc1
Genomic Map