Assay Details
| Gene Symbol: Adgrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174991.3 | NP_778156.2 | 15 - 16 | 3427 | 69 | |
| GenBank mRNA | AK147456.1 | - | 15 - 16 | 3412 | 69 | |
| AK147459.1 | - | 15 - 16 | 3380 | 69 | ||
| AK147494.1 | - | 15 - 16 | 3214 | 69 | ||
| AK147600.1 | - | 15 - 16 | 3189 | 69 | ||
| AK147607.1 | - | 15 - 16 | 3431 | 69 | ||
| AK220481.1 | - | 15 - 16 | 2806 | 69 | ||
| AY168408.1 | - | 15 - 16 | 2939 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adgrb1
Genomic Map