Assay Details
| Gene Symbol: Prickle3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290624.1 | NP_001277553.1 | 4 - 5 | 502 | 104 | |
| GenBank mRNA | AK037217.1 | - | 4 - 5 | 465 | 104 | |
| AK083704.1 | - | 3 - 4 | 469 | 104 | ||
| BC023671.1 | - | 4 - 5 | 498 | 104 | ||
| BC032173.1 | - | 1 - 2 | 823 | 104 | ||
| BC049258.1 | - | 4 - 5 | 444 | 5250488 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prickle3
Genomic Map