Assay Details
| Gene Symbol: Prickle3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290624.1 | NP_001277553.1 | 5 - 6 | 643 | 67 | |
| GenBank mRNA | AK037217.1 | - | 5 - 6 | 606 | 67 | |
| AK083704.1 | - | 4 - 5 | 610 | 67 | ||
| BC023671.1 | - | 5 - 6 | 639 | 67 | ||
| BC032173.1 | - | 2 - 3 | 964 | 67 | ||
| BC049258.1 | - | 5 - 6 | 585 | 5250488 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prickle3
Genomic Map