Assay Details
| Gene Symbol: Rae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175112.5 | NP_780321.1 | 7 - 8 | 796 | 83 | |
| GenBank mRNA | AK017746.1 | - | 7 - 8 | 84 | ||
| AK035278.1 | - | 7 - 8 | 742 | 83 | ||
| AK079383.1 | - | 7 - 8 | 777 | 83 | ||
| AK146058.1 | - | 7 - 8 | 761 | 83 | ||
| AK156681.1 | - | 7 - 8 | 761 | 83 | ||
| AK215169.1 | - | 1 - 2 | 57 | 83 | ||
| BC059051.1 | - | 7 - 8 | 718 | 6818771 | 83 | |
| BC060072.1 | - | 7 - 8 | 663 | 5718934 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rae1
Genomic Map