Assay Details
| Gene Symbol: Rae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175112.5 | NP_780321.1 | 8 - 9 | 905 | 83 | |
| GenBank mRNA | AK017746.1 | - | 8 - 9 | 827 | 82 | |
| AK035278.1 | - | 8 - 9 | 851 | 83 | ||
| AK079383.1 | - | 8 - 9 | 886 | 83 | ||
| AK146058.1 | - | 8 - 9 | 870 | 83 | ||
| AK156681.1 | - | 8 - 9 | 870 | 83 | ||
| AK178247.1 | - | 2 - 3 | 166 | 83 | ||
| AK206338.1 | - | 2 - 3 | 166 | 83 | ||
| AK215169.1 | - | 2 - 3 | 166 | 83 | ||
| BC059051.1 | - | 8 - 9 | 827 | 6818771 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rae1
Genomic Map