Assay Details
| Gene Symbol: Rae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175112.5 | NP_780321.1 | 10 - 11 | 1086 | 62 | |
| GenBank mRNA | AK014325.1 | - | 10 - 11 | 1042 | 62 | |
| AK017746.1 | - | 10 - 11 | 1007 | 62 | ||
| AK035278.1 | - | 10 - 11 | 1032 | 62 | ||
| AK079383.1 | - | 10 - 11 | 1067 | 62 | ||
| AK146058.1 | - | 10 - 11 | 1051 | 62 | ||
| AK156681.1 | - | 10 - 11 | 1051 | 62 | ||
| AK217457.1 | - | 1 - 2 | 73 | 62 | ||
| BC059051.1 | - | 10 - 11 | 1008 | 6818771 | 62 | |
| BC060072.1 | - | 8 - 9 | 3547 | 5718934 | 62 | |
| CO041965.1 | - | 2 - 3 | 62 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Rae1
Genomic Map