Assay Details
| Gene Symbol: Tmem161b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175187.5 | NP_780396.2 | 8 - 9 | 936 | 113 | |
| NR_029418.1 | 7 - 8 | 794 | 113 | |||
| GenBank mRNA | AK013305.1 | - | 7 - 8 | 765 | 113 | |
| AK088393.1 | - | 8 - 9 | 908 | 113 | ||
| BC037611.1 | - | 6 - 7 | 716 | 113 | ||
| BC051663.1 | - | 8 - 9 | 884 | 30020101 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem161b
Genomic Map