Assay Details
| Gene Symbol: Metap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175224.4 | NP_780433.1 | 3 - 4 | 293 | 71 | |
| GenBank mRNA | AK077694.1 | - | 3 - 4 | 294 | 71 | |
| AK152138.1 | - | 3 - 4 | 312 | 71 | ||
| AK153231.1 | - | 3 - 4 | 312 | 71 | ||
| AK153944.1 | - | 3 - 4 | 301 | 71 | ||
| AK165034.1 | - | 3 - 4 | 310 | 71 | ||
| BC096469.1 | - | 3 - 4 | 284 | 3603051 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Metap1
Genomic Map