Assay Details
| Gene Symbol: Metap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175224.4 | NP_780433.1 | 8 - 9 | 803 | 76 | |
| GenBank mRNA | AK077694.1 | - | 8 - 9 | 804 | 76 | |
| AK129054.1 | - | 3 - 4 | 2910 | 76 | ||
| AK152138.1 | - | 8 - 9 | 822 | 76 | ||
| AK153231.1 | - | 8 - 9 | 822 | 76 | ||
| AK153944.1 | - | 8 - 9 | 811 | 76 | ||
| BC027110.1 | - | 3 - 4 | 343 | 76 | ||
| BC096469.1 | - | 8 - 9 | 794 | 3603051 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Metap1
Genomic Map