Assay Details
| Gene Symbol: Metap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175224.4 | NP_780433.1 | 9 - 10 | 941 | 84 | |
| GenBank mRNA | AK006484.1 | - | 4 - 5 | 533 | 84 | |
| AK077694.1 | - | 9 - 10 | 942 | 84 | ||
| AK129054.1 | - | 4 - 5 | 3048 | 84 | ||
| AK152138.1 | - | 9 - 10 | 960 | 84 | ||
| AK153231.1 | - | 9 - 10 | 960 | 84 | ||
| BC027110.1 | - | 4 - 5 | 481 | 84 | ||
| BC096469.1 | - | 9 - 10 | 932 | 3603051 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Metap1
Genomic Map