Assay Details
| Gene Symbol: Arid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175251.4 | NP_780460.3 | 14 - 15 | 2090 | 59 | |
| GenBank mRNA | AK014481.1 | - | 1 - 2 | 54 | 59 | |
| CV559640.1 | - | 3 - 4 | 355 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014481.1 transcripts.
Set Membership: |
|

Arid2
Genomic Map