Assay Details
| Gene Symbol: Fzd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175284.3 | NP_780493.1 | 1 | 1360 | 61 | |
| GenBank mRNA | AF310970.1 | - | 1 | 1534 | 61 | |
| AK052950.1 | - | 1 | 1623 | 61 | ||
| AY509002.1 | - | 1 | 1360 | 61 | ||
| BC117759.2 | - | 1 | 1323 | 40090826 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fzd10
Genomic Map