Assay Details
Gene Symbol: Exoc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC025616.1 | - | 6 - 7 | 722 | 79 | |
BC026859.1 | - | 19 - 20 | 2101 | 2631201 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
