Assay Details
| Gene Symbol: Sephs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175400.6 | NP_780609.4 | 3 - 4 | 672 | 94 | |
| GenBank mRNA | AK044971.1 | - | 3 - 4 | 609 | 94 | |
| AK053638.1 | - | 3 - 4 | 707 | 94 | ||
| BC065165.1 | - | 3 - 4 | 633 | 6417054 | 94 | |
| BC066037.1 | - | 3 - 4 | 671 | 6405661 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sephs1
Genomic Map