Assay Details
| Gene Symbol: Sephs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175400.6 | NP_780609.4 | 4 - 5 | 780 | 91 | |
| GenBank mRNA | AK044971.1 | - | 4 - 5 | 717 | 91 | |
| AK053638.1 | - | 4 - 5 | 815 | 91 | ||
| BC065165.1 | - | 4 - 5 | 741 | 6417054 | 91 | |
| BC066037.1 | - | 4 - 5 | 779 | 6405661 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sephs1
Genomic Map