Assay Details
| Gene Symbol: Sephs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175400.6 | NP_780609.4 | 6 - 7 | 1033 | 85 | |
| GenBank mRNA | AK044971.1 | - | 6 - 7 | 970 | 85 | |
| AK047631.1 | - | 1 - 2 | 64 | 85 | ||
| AK053638.1 | - | 6 - 7 | 1068 | 85 | ||
| BC065165.1 | - | 6 - 7 | 994 | 6417054 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sephs1
Genomic Map