Assay Details
| Gene Symbol: Sephs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175400.6 | NP_780609.4 | 7 - 8 | 1127 | 76 | |
| GenBank mRNA | AK044971.1 | - | 7 - 8 | 1064 | 76 | |
| AK047631.1 | - | 2 - 3 | 158 | 76 | ||
| AK053638.1 | - | 7 - 8 | 1162 | 76 | ||
| BC066037.1 | - | 7 - 8 | 1126 | 6405661 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sephs1
Genomic Map