Assay Details
| Gene Symbol: Hcar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175520.4 | NP_780729.2 | 1 | 1273 | 82 | |
| GenBank mRNA | AK029064.1 | - | 1 | 1258 | 82 | |
| AK046700.1 | - | 1 | 1275 | 82 | ||
| BC141305.1 | - | 1 | 1144 | 9055917 | 82 | |
| EU809459.1 | - | 1 | 879 | 82 | ||
| EU809460.1 | - | 1 | 855 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P294059 | AK029064.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P294059 | AK046700.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P294059 | NM_175520.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P294059 | AK029064.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P294059 | AK046700.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P294059 | NM_175520.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044902 | BC141305.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044902 | EU809459.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044902 | EU809460.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044902 | AK029064.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044902 | AK046700.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 193891 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 663283 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 131792 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111085 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P294059 | AK029064.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P294059 | AK046700.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P294059 | NM_175520.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P294059 | AK029064.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P294059 | AK046700.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P294059 | NM_175520.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044902 | BC141305.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044902 | EU809459.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044902 | EU809460.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044902 | AK029064.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044902 | AK046700.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P294059 | AK029064.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P294059 | AK046700.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P294059 | NM_175520.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P294059 | AK029064.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P294059 | AK046700.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P294059 | NM_175520.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058094 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058074 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058065 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058084 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24058070 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111085 | - | - |
| Whole Mouse Genome G4122A | A_51_P294059 | AK029064.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P294059 | AK046700.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P294059 | NM_175520.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P294059 | AK029064.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P294059 | AK046700.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P294059 | NM_175520.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121278 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111085 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04453226 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04453213 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04453230 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04453205 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04453233 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04453220 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04453212 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04453204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24058059 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04453221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058065 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04948757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058097 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058054 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04397821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04453220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058103 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058096 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058077 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04453201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058046 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04453204 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058094 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058059 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058100 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04453235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04330045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04805950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058068 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24058056 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10533728 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533727 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533726 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438411_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10533725 | - | - |
| Mouse Genome 430 2.0 Array | 1438411_at | - | - |
| Mouse Genome 430 2.0 Array | 1438411_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 243270_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 243270_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500029236.mm.1 | TC0500003244.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500029236.mm.1 | NM_175520.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500029236.mm.1 | NM_175520.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500029235.mm.1 | TC0500003244.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500029235.mm.1 | NM_175520.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500029235.mm.1 | NM_175520.4 | Refseq |
| Mouse Expression Set 430 | 1438411_PM_at | - | - |
| Mouse Expression Set 430 | 1438411_at | - | - |
| Murine Genome U74v2 Set | 133937_PM_at | - | - |
| Murine Genome U74v2 Set | 133937_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438411_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438411_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 694319 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3190541 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3190541 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003190541 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2699134 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2699134 | - | - |
| mouse-6 v1.1 (Illumina) | scl26048.1.18_72-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3190541 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl26048.1.18_72-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3190541 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2699134 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2699134 | - | - |

Hcar1
Genomic Map