Assay Details
| Gene Symbol: Rab11fip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175543.3 | NP_780752.1 | 2 - 3 | 310 | 134 | |
| GenBank mRNA | AK046734.1 | - | 2 - 3 | 312 | 134 | |
| AK147219.1 | - | 2 - 3 | 381 | 134 | ||
| AK147344.1 | - | 2 - 3 | 507 | 134 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab11fip4
Genomic Map