Assay Details
| Gene Symbol: Oscar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175632.3 | NP_783440.1 | 1 - 2 | 137 | 126 | |
| GenBank mRNA | AF391161.1 | - | 1 - 2 | 113 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Oscar
Genomic Map