Assay Details
| Gene Symbol: Atp13a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284375.1 | NP_001271304.1 | 2 - 3 | 439 | 87 | |
| NM_175650.4 | NP_783581.2 | 2 - 3 | 439 | 87 | ||
| GenBank mRNA | AK083102.1 | - | 2 - 3 | 414 | 87 | |
| AK083121.1 | - | 2 - 3 | 440 | 87 | ||
| AK158355.1 | - | 2 - 3 | 259 | 87 | ||
| BC115739.1 | - | 2 - 3 | 374 | 40086065 | 87 | |
| CB524474.1 | - | 2 - 3 | 241 | 87 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC115739.1 transcripts.
Set Membership: |
|

Atp13a5
Genomic Map