Assay Details
| Gene Symbol: Bricd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175682.3 | NP_783613.1 | 3 - 4 | 614 | 81 | |
| GenBank mRNA | AK079437.1 | - | 3 - 4 | 615 | 81 | |
| BC096398.1 | - | 3 - 4 | 375 | 81 | ||
| BC119389.1 | - | 4 - 5 | 400 | 8734138 | 81 | |
| BC120670.1 | - | 4 - 5 | 400 | 40129593 | 81 | |
| BC145010.1 | - | 4 - 5 | 400 | 9053558 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1973164 | AK079437.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1973164 | BC096398.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1973164 | NM_175682.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1973164 | BC015279.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1973164 | AK088951.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170281 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1973164 | AK079437.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1973164 | BC096398.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1973164 | NM_175682.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1973164 | BC015279.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1973164 | AK088951.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31097525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31097531 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31097524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31097527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31097530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31097529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31097528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31097535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31097523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31097533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31097534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31097522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31097532 | - | - |
| Whole Mouse Genome G4122A | A_51_P143135 | AK079437.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143135 | NM_175682.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143135 | BC015279.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143135 | BC024545.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143135 | BC059106.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117167 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13293599 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13293586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31097584 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31097523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31097528 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31097527 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31097531 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13293577 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13293610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31097532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31097530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31097525 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31097534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31097524 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10442467 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442466 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442461 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442463 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442459 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442464 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442460 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442462 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10442458 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700002539.mm.1 | NM_175682.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002539.mm.1 | NM_175682.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002539.mm.1 | TC1700000340.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700002541.mm.1 | NM_175682.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002541.mm.1 | NM_175682.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002541.mm.1 | TC1700000340.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700002544.mm.1 | NM_175682.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002544.mm.1 | NM_175682.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002544.mm.1 | TC1700000340.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700002547.mm.1 | NM_175682.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002547.mm.1 | NM_175682.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002547.mm.1 | TC1700000340.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700002542.mm.1 | NM_175682.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002542.mm.1 | NM_175682.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002542.mm.1 | TC1700000340.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700002546.mm.1 | NM_175682.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002546.mm.1 | NM_175682.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002546.mm.1 | TC1700000340.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700002543.mm.1 | NM_175682.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002543.mm.1 | NM_175682.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002543.mm.1 | TC1700000340.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700002545.mm.1 | NM_175682.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002545.mm.1 | NM_175682.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002545.mm.1 | TC1700000340.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 134023_PM_at | - | - |
| Murine Genome U74v2 Set | 134023_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 491057 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1980148 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1980148 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001980148 | - | - |
| mouse-6 v1.1 (Illumina) | scl51014.5.1_34-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1980148 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl51014.5.1_34-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1980148 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2951995 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2951995 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2764325 | - | - |

Bricd5
Genomic Map