Assay Details
| Gene Symbol: Zfp608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175751.4 | NP_786927.2 | 5 - 6 | 4245 | 79 | |
| GenBank mRNA | AK076178.1 | - | 1 - 2 | 1121 | 79 | |
| AK077030.1 | - | 4 - 5 | 2972 | 79 | ||
| BC059268.1 | - | 4 - 5 | 3912 | 79 | ||
| BC092219.1 | - | 5 - 6 | 3978 | 6830664 | 79 | |
| BC151073.1 | - | 4 - 5 | 3764 | 9087986 | 79 | |
| BC151075.1 | - | 4 - 5 | 3764 | 9087988 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp608
Genomic Map