Assay Details
| Gene Symbol: Grik4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175481.5 | NP_780690.2 | 8 - 9 | 944 | 76 | |
| GenBank mRNA | AK032029.1 | - | 8 - 9 | 945 | 76 | |
| AK080582.1 | - | 8 - 9 | 1072 | 76 | ||
| BC118010.2 | - | 7 - 8 | 859 | 40096690 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Grik4
Genomic Map