Assay Details
| Gene Symbol: Cpe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013494.3 | NP_038522.2 | 6 - 7 | 1200 | 92 | |
| GenBank mRNA | AK013509.1 | - | 6 - 7 | 896 | 92 | |
| AK047891.1 | - | 6 - 7 | 1191 | 92 | ||
| AK190413.1 | - | 1 - 2 | 61 | 92 | ||
| AK192422.1 | - | 1 - 2 | 61 | 92 | ||
| AK193438.1 | - | 1 - 2 | 61 | 92 | ||
| AK194077.1 | - | 1 - 2 | 61 | 92 | ||
| AK194100.1 | - | 1 - 2 | 61 | 92 | ||
| AK202236.1 | - | 1 - 2 | 61 | 92 | ||
| AK212757.1 | - | 1 - 2 | 61 | 92 | ||
| BC010197.1 | - | 6 - 7 | 1200 | 3157558 | 92 | |
| U23184.1 | - | 6 - 7 | 1109 | 92 | ||
| X61232.1 | - | 6 - 7 | 92 | |||
More Information
Set Membership: |
|

Cpe
Genomic Map