Assay Details
| Gene Symbol: Npr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173788.3 | NP_776149.1 | 17 - 18 | 2894 | 93 | |
| GenBank mRNA | AK158195.1 | - | 12 - 13 | 1445 | 93 | |
| BC023420.1 | - | 4 - 5 | 535 | 93 | ||
| BC037458.1 | - | 12 - 13 | 1350 | 93 | ||
| BC042470.1 | - | 17 - 18 | 2894 | 5137261 | 93 | |
| BC049896.1 | - | 7 - 8 | 989 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npr2
Genomic Map