| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2085370 |
AK080607.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2085370 |
NM_001172114.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2085370 |
NM_153792.2 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2085370 |
NM_001172113.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P448870 |
AK043912.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P448870 |
BC094931.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P448870 |
NM_177375.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P448870 |
AK043912.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P448870 |
BC094931.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P448870 |
NM_177375.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
527817 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
195387 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
655753 |
D85818.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
655753 |
NM_011293.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
136966 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P159958 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P448870 |
AK043912.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P448870 |
BC094931.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P448870 |
NM_177375.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P448870 |
AK043912.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P448870 |
BC094931.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P448870 |
NM_177375.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P179342 |
AK080607.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P448870 |
AK043912.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P448870 |
BC094931.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P448870 |
NM_177375.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P448870 |
AK043912.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P448870 |
BC094931.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P448870 |
NM_177375.1 |
mouse_transcriptome |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31097979 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13324380 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31097978 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13324374 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13540383 |
- |
- |