Assay Details
| Gene Symbol: Crygs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009967.2 | NP_034097.1 | 2 - 3 | 318 | 85 | |
| GenBank mRNA | AF032995.1 | - | 2 - 3 | 314 | 85 | |
| AK143060.1 | - | 2 - 3 | 315 | 85 | ||
| BC056452.1 | - | 2 - 3 | 294 | 6824006 | 85 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1975315 | NM_009967.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 390856 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 912016 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 246999 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 95399 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145200 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1975315 | NM_009967.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | ERCC-00168_248 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P459498 | NM_009967.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P459498 | AF032995.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P459498 | AK143060.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599968 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599983 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599981 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30599970 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08532546 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145200 | - | - |
| Whole Mouse Genome G4122A | A_51_P459498 | NM_009967.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P459498 | AF032995.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P459498 | AK143060.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135784 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145200 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12695935 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12695947 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12695940 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12731378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30599919 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12992000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599914 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599871 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599880 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12822951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13035080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12695941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599978 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599972 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12695932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12822946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12991983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599919 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12496811 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599929 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599916 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12850664 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12496798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12808070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12928373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599976 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599963 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13035077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12695940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599899 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12860988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12991993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12822949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12731378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12850667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30599975 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12982470 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12822939 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10438670 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438671 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438669 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420453_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10438668 | - | - |
| Mouse Genome 430 2.0 Array | 1420453_at | - | - |
| Mouse Genome 430 2.0 Array | 1420453_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12970_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12970_at | - | - |
| Mouse Expression Set 430 | 1420453_at | - | - |
| Mouse Expression Set 430 | 1420453_PM_at | - | - |
| Murine Genome U74v2 Set | 93391_at | - | - |
| Murine Genome U74v2 Set | 93391_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420453_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420453_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420453_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420453_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 402761 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2680398 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2680398 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002680398 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2702266 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2702266 | - | - |
| mouse-6 v1.1 (Illumina) | scl48638.3.1_115-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2680398 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl48638.3.1_115-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2680398 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2702266 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2702266 | - | - |

Crygs
Genomic Map