Assay Details
| Gene Symbol: Gart | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010256.2 | NP_034386.2 | 7 - 8 | 856 | 111 | |
| GenBank mRNA | AK146355.1 | - | 7 - 8 | 841 | 111 | |
| AK146478.1 | - | 7 - 8 | 794 | 111 | ||
| AK168445.1 | - | 7 - 8 | 806 | 111 | ||
| AK168458.1 | - | 7 - 8 | 816 | 111 | ||
| AK168501.1 | - | 7 - 8 | 819 | 111 | ||
| AK168724.1 | - | 7 - 8 | 831 | 111 | ||
| AK168796.1 | - | 7 - 8 | 831 | 111 | ||
| AK168864.1 | - | 7 - 8 | 807 | 111 | ||
| AK168876.1 | - | 7 - 8 | 808 | 111 | ||
| BC070465.1 | - | 7 - 8 | 803 | 30611835 | 111 | |
| U01023.1 | - | 7 - 8 | 827 | 111 | ||
| U01024.1 | - | 6 - 7 | 761 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gart
Genomic Map