Assay Details
| Gene Symbol: Hexa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010421.4 | NP_034551.2 | 10 - 11 | 1207 | 67 | |
| GenBank mRNA | AK075895.1 | - | 10 - 11 | 1183 | 67 | |
| AK075911.1 | - | 10 - 11 | 1208 | 67 | ||
| AK080777.1 | - | 10 - 11 | 1180 | 67 | ||
| AK144168.1 | - | 10 - 11 | 1335 | 67 | ||
| AK150444.1 | - | 10 - 11 | 1178 | 67 | ||
| AK151960.1 | - | 10 - 11 | 1182 | 67 | ||
| AK159091.1 | - | 11 - 12 | 1178 | 67 | ||
| AK159814.1 | - | 10 - 11 | 1193 | 67 | ||
| AK159890.1 | - | 10 - 11 | 1180 | 67 | ||
| AK160112.1 | - | 10 - 11 | 1167 | 67 | ||
| AK167013.1 | - | 10 - 11 | 1193 | 67 | ||
| AK168186.1 | - | 10 - 11 | 1207 | 67 | ||
| AK170903.1 | - | 10 - 11 | 1194 | 67 | ||
| BC010755.1 | - | 10 - 11 | 1176 | 4225233 | 67 | |
| U07631.1 | - | 10 - 11 | 1199 | 67 | ||
| X64331.1 | - | 10 - 11 | 1150 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hexa
Genomic Map