Assay Details
| Gene Symbol: Hexa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010421.4 | NP_034551.2 | 5 - 6 | 626 | 77 | |
| GenBank mRNA | AK075895.1 | - | 5 - 6 | 602 | 77 | |
| AK075911.1 | - | 5 - 6 | 627 | 77 | ||
| AK080777.1 | - | 5 - 6 | 599 | 77 | ||
| AK144168.1 | - | 5 - 6 | 754 | 77 | ||
| AK150444.1 | - | 5 - 6 | 597 | 77 | ||
| AK151960.1 | - | 5 - 6 | 601 | 77 | ||
| AK159091.1 | - | 5 - 6 | 618 | 77 | ||
| AK159814.1 | - | 5 - 6 | 612 | 77 | ||
| AK159890.1 | - | 5 - 6 | 599 | 77 | ||
| AK160112.1 | - | 5 - 6 | 586 | 77 | ||
| AK167013.1 | - | 5 - 6 | 612 | 77 | ||
| AK168186.1 | - | 5 - 6 | 626 | 77 | ||
| AK170903.1 | - | 5 - 6 | 613 | 77 | ||
| BC010755.1 | - | 5 - 6 | 595 | 4225233 | 77 | |
| U07631.1 | - | 5 - 6 | 618 | 77 | ||
| X64331.1 | - | 5 - 6 | 569 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hexa
Genomic Map